2-Brom-4-isopropylphenylisothiocyanat

Lieferant: Thermo Scientific

SB01968.1 SB01968.10
MAYBSB01968.1EA 0 EUR
MAYBSB01968.1 MAYBSB01968.10
2-Brom-4-isopropylphenylisothiocyanat
2-Brom-4-isopropylphenylisothiocyanat
Formel: C₁₀H₁₀BrNS
Molecular Weight: 256,17 g/mol
CAS-Nummer: 246166-33-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR