2,6-Dibrom-4-isopropylanilin
Lieferant: Thermo Scientific
|
Achtung
|
BTB04550.1
BTB04550.10
BTB04550.50
MAYBBTB04550.1EA
0
EUR
MAYBBTB04550.1
MAYBBTB04550.10
MAYBBTB04550.50
2,6-Dibrom-4-isopropylanilin
2,6-Dibrom-4-isopropylanilin
Formel:
C₉H₁₁Br₂N Siedepunkt: 316 °C (1013 hPa) Schmelzpunkt: 45…47 °C |
MDL:
MFCD00015475 CAS-Nummer: 10546-65-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...