2,6-Dibrom-4-propylanilin

Lieferant: CHEMPUR
Achtung

BTB04637.250 BTB04637.1
CHMPBTB04637.250EA 0 EUR
CHMPBTB04637.250 MAYBBTB04637.1
2,6-Dibrom-4-propylanilin
2,6-Dibrom-4-propylanilin
Formel: C₉H₁₁Br₂N
Molecular Weight: 293 g/mol
Siedepunkt: 324 °C (1013 hPa)
Schmelzpunkt: 55…57 °C
MDL: MFCD00051706
CAS-Nummer: 10546-64-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR