3-Brom-p-toluidin ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3-Brom-4-methylanilin, 4-Amino-2-bromtoluol

B24670.06 B24670.14
B24670.06EA 46.4 EUR
B24670.06 B24670.14
3-Brom-p-toluidin ≥97%
3-Brom-p-toluidin

Purity: 97% CAS No.: 7745-91-7 Molecular Formula: C7H8BrN Formula Weight: 186.06 Flash Point: 110°(230°F) Refractive Index: 1.6115 Odour: Characteristic Melting Point: 25-28° Boiling Point: 254-257° Beilstein Reference: 1562057 UN No.: 2811 MDL No.: MFCD00134176

Formel: C₇H₈BrN
Molecular Weight: 186,05 g/mol
Siedepunkt: 254…257 °C
Schmelzpunkt: 25…28 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00134176
CAS-Nummer: 7745-91-7
EINECS: 231-807-8
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR