4-Brom-m-toluidin ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 4-Brom-3-methylanilin, 3-Amino-6-bromtoluol, 5-Amino-2-bromtoluol

A15417.09 A15417.18
A15417.09EA 23.8 EUR
A15417.09 A15417.18
4-Brom-m-toluidin ≥97%
4-Brom-m-toluidin

Purity: 97% CAS No.: 6933-10-4 Molecular Formula: C7H8BrN Formula Weight: 186.06 Odour: Characteristic Melting Point: 79-83° Boiling Point: 239-241° Beilstein Reference: 2689600 UN No.: 2811 MDL No.: MFCD00007828

Formel: C₇H₈BrN
Molecular Weight: 186,05 g/mol
Siedepunkt: 239…241 °C
Schmelzpunkt: 79…83 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007828
CAS-Nummer: 6933-10-4
EINECS: 230-056-3
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR