5-Brom-o-kresol 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Brom-2-methylphenol, 4-Brom-2-hydroxytoluol, 1-Brom-3-hydroxy-4-methylbenzol

H26956.09 H26956.03
H26956.09EA 184 EUR
H26956.09 H26956.03
5-Brom-o-kresol 95%
5-Brom-o-kresol

Purity: 95% CAS No.: 36138-76-8 Molecular Formula: C7H7BrO Formula Weight: 187.04 Melting Point: 77-80° MDL No.: MFCD08061906

Formel: C₇H₇BrO
Molecular Weight: 187,04 g/mol
Schmelzpunkt: 77…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08061906
CAS-Nummer: 36138-76-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR