4-Brombenzylalkohol ≥99%

Lieferant: Thermo Scientific

Synonyme: (4-Bromophenyl)methanol

A13197.09 A13197.18
A13197.09EA 43.4 EUR
A13197.09 A13197.18
4-Brombenzylalkohol ≥99%
4-Brombenzylalkohol

Purity: 99% CAS No.: 873-75-6 Molecular Formula: C7H7BrO Formula Weight: 187.04 Melting Point: 76-79° Beilstein Reference: 1931620

Formel: C₇H₇BrO
Molecular Weight: 187,04 g/mol
Siedepunkt: 268 °C (1013 hPa)
Schmelzpunkt: 76…79 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004650
CAS-Nummer: 873-75-6
EINECS: 212-851-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR