3',4'-Dichloracetophenon ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
1-(3,4-dichlorophenyl)ethanone
A13568.14
A13568.22
A13568.14EA
68.7
EUR
A13568.14
A13568.22
3',4'-Dichloracetophenon ≥98%
3',4'-Dichloracetophenon
Purity: 98+%
CAS No.: 2642-63-9
Molecular Formula: C8H6Cl2O
Formula Weight: 189.04
Melting Point: 72-76°
Boiling Point: 134-135mm/12mm
MDL No.: MFCD00000553
Formel:
C₈H₆Cl₂O Molecular Weight: 189.04 g/mol Siedepunkt: 134…135°C (12 mmHg) Schmelzpunkt: 72…76 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000553 CAS-Nummer: 2642-63-9 EINECS: 220-146-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...