2',6'-Dichloracetophenon

Lieferant: FLUOROCHEM
Achtung

Synonyme: 1-(2,6-Dichlorophenyl)ethanone

009766-1G 009766-5G
FLUO009766-1GEA 31.1 EUR
FLUO009766-1G FLUO009766-5G
2',6'-Dichloracetophenon
2',6'-Dichloracetophenon
Formel: C₈H₆Cl₂O
Molecular Weight: 189,04 g/mol
MDL: MFCD00016339
CAS-Nummer: 2040-05-3
EINECS: 218-035-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR