4'-Chlorpropiophenon ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: p-Chlorpropiophenon

A11633.14 A11633.22
A11633.14EA 35.9 EUR
A11633.14 A11633.22
4'-Chlorpropiophenon ≥98%
4'-Chlorpropiophenon

Purity: 98% CAS No.: 6285-05-8 Molecular Formula: C9H9ClO Formula Weight: 168.62 Flash Point: 110°(230°F) Melting Point: 33-36° Boiling Point: 125-126°/13mm Solubility: Insoluble in water Beilstein Reference: 1100638 MDL No.: MFCD00000626

Formel: C₉H₉ClO
Molecular Weight: 168,62 g/mol
Siedepunkt: 125…126 °C (13 mmHg)
Schmelzpunkt: 33…36 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000626
CAS-Nummer: 6285-05-8
EINECS: 228-511-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR