2-Chlor-6-methylbenzamid

Lieferant: CHEMPUR

Synonyme: 2-Chlor-6-methylbenzoesäureamid

TL00181.1 TL00181.10
CHMPTL00181.1EA 0 EUR
CHMPTL00181.1 CHMPTL00181.10
2-Chlor-6-methylbenzamid
2-Chlor-6-methylbenzamid
Formel: C₈H₈ClNO
Molecular Weight: 169,61 g/mol
MDL: MFCD00052820
CAS-Nummer: 101080-58-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR