6-Chlor-o-toluylsäure

Lieferant: FLUOROCHEM
Achtung

Synonyme: 2-Chloro-6-methylbenzoic acid

018392-1G 018392-5G
FLUO018392-1GEA 31.1 EUR
FLUO018392-1G FLUO018392-5G
6-Chlor-o-toluylsäure
6-Chlor-o-toluylsäure
Formel: C₈H₇ClO₂
Molecular Weight: 170.6 g/mol
MDL: MFCD00045799
CAS-Nummer: 21327-86-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR