4-Chlor-m-toluylsäure

Lieferant: FLUOROCHEM
Achtung

Synonyme: 4-Chloro-3-methylbenzoic acid

018394-10G 018394-25G
FLUO018394-10GEA 31.1 EUR
FLUO018394-10G FLUO018394-25G
4-Chlor-m-toluylsäure
4-Chlor-m-toluylsäure
Formel: C₈H₇ClO₂
Molecular Weight: 170.6 g/mol
MDL: MFCD00045853
CAS-Nummer: 7697-29-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR