α-Chlor-3,4-difluortoluol
Lieferant: FLUOROCHEM
|
Gefahr
|
Synonyme:
1-(Chlormethyl)-3,4-difluorbenzol, 3,4-Difluorbenzylchlorid
007846-5G
007846-25G
FLUO007846-5GEA
31.1
EUR
FLUO007846-5G
FLUO007846-25G
α-Chlor-3,4-difluortoluol
α-Chlor-3,4-difluortoluol
Formel:
C₇H₅ClF₂ Molecular Weight: 162.57 g/mol |
MDL:
MFCD00236218 CAS-Nummer: 698-80-6 UN: 1760 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...