2-Chlor-5-(trifluormethyl)benzoylchlorid
Lieferant: FLUOROCHEM
005712-5G
005712-1G
FLUO005712-5GEA
146
EUR
FLUO005712-5G
FLUO005712-1G
2-Chlor-5-(trifluormethyl)benzoylchlorid
2-Chlor-5-(trifluormethyl)benzoylchlorid
Formel:
C₈H₃Cl₂F₃O Molecular Weight: 243,01 g/mol Siedepunkt: 59…61 °C (1013 hPa) |
MDL:
MFCD01631634 CAS-Nummer: 657-05-6 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...