2-Chlor-4-(trifluormethyl)anilin

Lieferant: FLUOROCHEM
Achtung

Synonyme: 4-Amino-3-chlor-α,α,α-trifluortoluol, 2-Chlor-α,α,α-trifluor-p-toluidin

003965-100G 003965-500G 003965-25G
FLUO003965-100GEA 152 EUR
FLUO003965-100G FLUO003965-500G FLUO003965-25G
2-Chlor-4-(trifluormethyl)anilin
2-Chlor-4-(trifluormethyl)anilin
Formel: C₇H₅ClF₃N
Molecular Weight: 195,57 g/mol
Siedepunkt: 89 °C (0,5 mmHg)
MDL: MFCD00042563
CAS-Nummer: 39885-50-2
EINECS: 254-674-8
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR