2,4-Dichlor-5-nitrobenzotrifluorid

Lieferant: FLUOROCHEM
Achtung

Synonyme: 2,4-Dichloro-5-nitrobenzotrifluoride, 1,3-Dichloro-5-nitro-4-(trifluoromethyl)benzene

002827-5G 002827-25G 002827-100G
FLUO002827-5GEA 0 EUR
FLUO002827-5G FLUO002827-25G FLUO002827-100G
2,4-Dichlor-5-nitrobenzotrifluorid
2,4-Dichlor-5-nitrobenzotrifluorid
Formel: C₇H₂Cl₂F₃NO₂
Molecular Weight: 260 g/mol
Schmelzpunkt: 59,5 °C
MDL: MFCD00007073
CAS-Nummer: 400-70-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR