3-Chlor-4-nitrobenzotrifluorid
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
2-Chlor-1-nitro-4-(trifluormethyl)benzol, 3-Chlor-4-nitrobenzotrifluorid, 1-Chlor-2-nitro-5-(trifluormethyl)benzol
009097-1G
009097-5G
FLUO009097-1GEA
83.9
EUR
FLUO009097-1G
FLUO009097-5G
3-Chlor-4-nitrobenzotrifluorid
3-Chlor-4-nitrobenzotrifluorid
Formel:
C₇H₃ClF₃NO₂ |
MDL:
MFCD00272564 CAS-Nummer: 402-11-9 EINECS: 206-936-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...