3-Chlor-2,6-difluorbenzylalkohol

Lieferant: FLUOROCHEM

Synonyme: (3-Chloro-2,6-difluorophenyl)methanol

005597-5G 005597-1G
FLUO005597-5GEA 0 EUR
FLUO005597-5G FLUO005597-1G
3-Chlor-2,6-difluorbenzylalkohol
3-Chlor-2,6-difluorbenzylalkohol
Formel: C₇H₅ClF₂O
Molecular Weight: 178.57 g/mol
MDL: MFCD01631325
CAS-Nummer: 252004-35-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR