2-Chlor-5-fluorbenzylalkohol

Lieferant: FLUOROCHEM

005624-5G 005624-10G 005624-1G
FLUO005624-5GEA 79.1 EUR
FLUO005624-5G FLUO005624-10G FLUO005624-1G
2-Chlor-5-fluorbenzylalkohol
2-Chlor-5-fluorbenzylalkohol
Formel: C₇H₆ClFO
Molecular Weight: 160,58 g/mol
MDL: MFCD01631418
CAS-Nummer: 261762-59-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR