5-Chlor-2-fluorbenzylalkohol

Lieferant: FLUOROCHEM

005627-1G 005627-5G
FLUO005627-1GEA 51.8 EUR
FLUO005627-1G FLUO005627-5G
5-Chlor-2-fluorbenzylalkohol
5-Chlor-2-fluorbenzylalkohol
Formel: C₇H₆ClFO
Molecular Weight: 160,58 g/mol
MDL: MFCD01631436
CAS-Nummer: 188723-58-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR