3-Chlor-4-iodtoluol
Lieferant: FLUOROCHEM
018384-5G
018384-1G
FLUO018384-5GEA
67.8
EUR
FLUO018384-5G
FLUO018384-1G
3-Chlor-4-iodtoluol
3-Chlor-4-iodtoluol
Formel:
C₇H₆ClI Molecular Weight: 252.48 g/mol |
MDL:
MFCD00079734 CAS-Nummer: 116632-42-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...