5-Chlor-2-iodtoluol

Lieferant: FLUOROCHEM

016386-10G 016386-25G
FLUO016386-10GEA 0 EUR
FLUO016386-10G FLUO016386-25G
5-Chlor-2-iodtoluol
5-Chlor-2-iodtoluol
Formel: C₇H₆ClI
Molecular Weight: 252.48 g/mol
MDL: MFCD00060665
CAS-Nummer: 23399-70-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR