2-Chlor-4-methylphenylisothiocyanat
Lieferant: FLUOROCHEM
008763-5G
008763-1G
FLUO008763-5GEA
116
EUR
FLUO008763-5G
FLUO008763-1G
2-Chlor-4-methylphenylisothiocyanat
2-Chlor-4-methylphenylisothiocyanat
Formel:
C₈H₆ClNS Molecular Weight: 183.66 g/mol |
MDL:
MFCD00046791 CAS-Nummer: 57878-93-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...