3-Chlor-2-methylphenylisothiocyanat

Lieferant: FLUOROCHEM

Synonyme: 1-Chlor-3-isothiocyanato-2-methylbenzol

008765-5G 008765-10G
FLUO008765-5GEA 45.9 EUR
FLUO008765-5G FLUO008765-10G
3-Chlor-2-methylphenylisothiocyanat
3-Chlor-2-methylphenylisothiocyanat
Formel: C₈H₆ClNS
Molecular Weight: 183,66 g/mol
MDL: MFCD00022056
CAS-Nummer: 19241-35-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR