5-Chlor-2-methylphenylisothiocyanat
Lieferant: Thermo Scientific
Synonyme:
1-Chloro-3-isothiocyanato-4-methylbenzene
SB01664.1
SB01664.10
SB01664.50
MAYBSB01664.1EA
0
EUR
MAYBSB01664.1
MAYBSB01664.10
MAYBSB01664.50
5-Chlor-2-methylphenylisothiocyanat
5-Chlor-2-methylphenylisothiocyanat
Formel:
C₈H₆ClNS Molecular Weight: 183,66 g/mol Siedepunkt: 268 °C (1013 hPa) Dichte: 1,248 g/cm³ (20 °C) |
MDL:
MFCD00046801 CAS-Nummer: 19241-36-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...