5-Chlor-2-methylphenylisothiocyanat
Lieferant: FLUOROCHEM
Synonyme:
1-Chloro-3-isothiocyanato-4-methylbenzene
008768-5G
008768-1G
FLUO008768-5GEA
55.4
EUR
FLUO008768-5G
FLUO008768-1G
5-Chlor-2-methylphenylisothiocyanat
5-Chlor-2-methylphenylisothiocyanat
Formel:
C₈H₆ClNS Molecular Weight: 183,66 g/mol Siedepunkt: 268 °C (1013 hPa) Dichte: 1,248 g/cm³ (20 °C) |
MDL:
MFCD00046801 CAS-Nummer: 19241-36-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...