4-Chlor-2-methylphenylisothiocyanat
Lieferant: FLUOROCHEM
008767-1G
008767-5G
008767-25G
FLUO008767-1GEA
32.4
EUR
FLUO008767-1G
FLUO008767-5G
FLUO008767-25G
4-Chlor-2-methylphenylisothiocyanat
4-Chlor-2-methylphenylisothiocyanat
Formel:
C₈H₆ClNS Molecular Weight: 183,66 g/mol Siedepunkt: 136 °C (13 mmHg) Schmelzpunkt: 36 °C |
MDL:
MFCD00041089 CAS-Nummer: 23165-53-9 EINECS: 245-472-0 UN: 2923 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...