3-Chlorbenzylamin ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(3-Chlorophenyl)methanamine
A14078.09
A14078.18
A14078.09EA
86.1
EUR
A14078.09
A14078.18
3-Chlorbenzylamin ≥97%
3-Chlorbenzylamin
Purity: 97+%
CAS No.: 4152-90-3
Molecular Formula: C7H8ClN
Formula Weight: 141.60
Flash Point: 98°(208°F)
Refractive Index: 1.5590
Odour: Amine-like
Boiling Point: 226°
Beilstein Reference: 774509
UN No.: 2735
MDL No.: MFCD00040752
Formel:
C₇H₈ClN Molecular Weight: 141,6 g/mol Siedepunkt: 226 °C Dichte: 1,159 Flammpunkt: 98 °C (208 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040752 CAS-Nummer: 4152-90-3 EINECS: 223-985-0 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...