3-Chlorbenzylamin
Lieferant: FLUOROCHEM
|
Gefahr
|
Synonyme:
(3-Chlorophenyl)methanamine
008869-5G
008869-25G
FLUO008869-5GEA
48.8
EUR
FLUO008869-5G
FLUO008869-25G
3-Chlorbenzylamin
3-Chlorbenzylamin
Formel:
C₇H₈ClN Molecular Weight: 141,6 g/mol Siedepunkt: 40 °C (0,8 mmHg) Dichte: 1,159 g/cm³ (20 °C) |
MDL:
MFCD00040752 CAS-Nummer: 4152-90-3 EINECS: 223-985-0 UN: 2735 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...