6-Chlor-m-toluidin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
H26209.06
H26209.14
H26209.06EA
93.4
EUR
H26209.06
H26209.14
6-Chlor-m-toluidin ≥98%
6-Chlor-m-toluidin
Purity: 98%
CAS No.: 95-81-8
Molecular Formula: C7H8ClN
Formula Weight: 141.60
Flash Point: 107°(225°F)
Melting Point: 31-33°
Boiling Point: 228-230°
UN No.: 2239
MDL No.: MFCD00007674
Formel:
C₇H₈ClN Molecular Weight: 141,6 g/mol Siedepunkt: 228…230 °C Schmelzpunkt: 31…33 °C Flammpunkt: 107 °C (225 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007674 CAS-Nummer: 95-81-8 EINECS: 202-454-7 UN: 2239 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...