4-Chlor-m-toluidin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
5-Amino-2-chlortoluol, 4-Chlor-3-methylanilin
B22234.06
B22234.14
B22234.06EA
59.3
EUR
B22234.06
B22234.14
4-Chlor-m-toluidin ≥98%
4-Chlor-m-toluidin
Purity: 98%
CAS No.: 7149-75-9
Molecular Formula: C7H8ClN
Formula Weight: 141.60
Melting Point: 82-85°
Beilstein Reference: 636522
UN No.: 2239
MDL No.: MFCD00066332
Formel:
C₇H₈ClN Molecular Weight: 141,6 g/mol Siedepunkt: 241 °C (1013 hPa) Schmelzpunkt: 82…85 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066332 CAS-Nummer: 7149-75-9 UN: 2239 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...