2-Chlor-5-nitrobenzylalkohol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (2-Chloro-5-nitrophenyl)methanol

A14205.22 A14205.14 A14205.06
A14205.22EA 278 EUR
A14205.22 A14205.14 A14205.06
2-Chlor-5-nitrobenzylalkohol ≥98%
2-Chlor-5-nitrobenzylalkohol

Purity: 98% CAS No.: 80866-80-4 Molecular Formula: C7H6ClNO3 Formula Weight: 187.58 Melting Point: 75-79° Beilstein Reference: 3248759

Formel: C₇H₆ClNO₃
Molecular Weight: 187.58 g/mol
Schmelzpunkt: 75…79 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007297
CAS-Nummer: 80866-80-4
EINECS: 279-584-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR