3-Chlor-2-nitrobenzylalkohol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

647586-1G 647586-5G
SIAL647586-1GEA 60.4 EUR
SIAL647586-1G SIAL647586-5G
3-Chlor-2-nitrobenzylalkohol, Sigma-Aldrich®
3-Chlor-2-nitrobenzylalkohol
Formel: C₇H₆ClNO₃
Molecular Weight: 187.58 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04038814
CAS-Nummer: 77158-86-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR