4-Chlor-3-nitrobenzylalkohol ≥98%

Lieferant: Thermo Scientific

A16024.06 A16024.14
A16024.06EA 34 EUR
A16024.06 A16024.14
4-Chlor-3-nitrobenzylalkohol ≥98%
4-Chlor-3-nitrobenzylalkohol

Purity: 98% CAS No.: 55912-20-4 Molecular Formula: C7H6ClNO3 Formula Weight: 187.58 Melting Point: 62-64° MDL No.: MFCD00007086

Formel: C₇H₆ClNO₃
Molecular Weight: 187,58 g/mol
Schmelzpunkt: 62…64 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007086
CAS-Nummer: 55912-20-4
EINECS: 259-901-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR