α-Chlor-3,5-dinitrotoluol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,5-Dinitrobenzyl chloride

M-Clarity™ Qualitätsstufe = MQ200

191647-10G 191647-50G
SIAL191647-10GEA 196 EUR
SIAL191647-10G SIAL191647-50G
α-Chlor-3,5-dinitrotoluol, Sigma-Aldrich®
α-Chlor-3,5-dinitrotoluol
Formel: C₇H₅ClN₂O₄
Molecular Weight: 216.58 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 74367-78-5
EINECS: 277-841-7
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR