2-Chlor-5-(trifluormethyl)anisol

Lieferant: FLUOROCHEM

Synonyme: 1-Chloro-2-methoxy-4-(trifluoromethyl)benzene

005705-1G 005705-5G
FLUO005705-1GEA 58.1 EUR
FLUO005705-1G FLUO005705-5G
2-Chlor-5-(trifluormethyl)anisol
2-Chlor-5-(trifluormethyl)anisol
Formel: C₈H₆ClF₃O
Molecular Weight: 210.58 g/mol
MDL: MFCD01631470
CAS-Nummer: 402-08-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR