4-Chlor-3-(trifluormethyl)anisol

Lieferant: FLUOROCHEM
Gefahr

005706-5G 005706.10G 005706-1G
FLUO005706-5GEA 188 EUR
FLUO005706-5G FLUO005706.10G FLUO005706-1G
4-Chlor-3-(trifluormethyl)anisol
4-Chlor-3-(trifluormethyl)anisol
Formel: C₈H₆ClF₃O
Molecular Weight: 210.58 g/mol
MDL: MFCD01631404
CAS-Nummer: 400-73-7
UN: 3272
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR