3,4,5-Trimethoxybenzylchlorid, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: 5-(Chlormethyl)-1,2,3-trimethoxybenzol

92176-25G 92176-5G
SIAL92176-25GEA 369 EUR
SIAL92176-25G SIAL92176-5G
3,4,5-Trimethoxybenzylchlorid, Sigma-Aldrich®
3,4,5-Trimethoxybenzylchlorid
Formel: C₁₀H₁₃ClO₃
Molecular Weight: 216,66 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00040716
CAS-Nummer: 3840-30-0
EINECS: 223-330-9
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR