2-Chlorbenzylalkohol 99%
Lieferant: Thermo Fisher Scientific
Synonyme:
(2-Chlorophenyl)methanol
163420100
163420500
ACRO163420100EA
23.5
EUR
ACRO163420100
ACRO163420500
2-Chlorbenzylalkohol 99%
2-Chlorbenzylalkohol
Formel:
C₇H₇ClO Molecular Weight: 142.58 g/mol Siedepunkt: 227 Schmelzpunkt: 69…72 °C |
MDL:
MFCD00004604 CAS-Nummer: 17849-38-6 EINECS: 241-801-7 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 69 to 72 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...