2-Chlorbenzylalkohol ≥98%

Lieferant: Thermo Scientific

Synonyme: (2-Chlorophenyl)methanol

A13657.14 A13657.22
A13657.14EA 41.9 EUR
A13657.14 A13657.22
2-Chlorbenzylalkohol ≥98%
2-Chlorbenzylalkohol

Purity: 98+% CAS No.: 17849-38-6 Molecular Formula: C7H7ClO Formula Weight: 142.59 Melting Point: 69-71° Boiling Point: 227° MDL No.: MFCD00004604

Formel: C₇H₇ClO
Molecular Weight: 142,58 g/mol
Siedepunkt: 227 °C
Schmelzpunkt: 69…71 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004604
CAS-Nummer: 17849-38-6
EINECS: 241-801-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR