3-Chlorbenzylalkohol

Lieferant: CHEMPUR

Synonyme: (3-Chlorophenyl)methanol

FL1376.25 FL1376.100
CHMPFL1376.25EA 0 EUR
CHMPFL1376.25 CHMPFL1376.100
3-Chlorbenzylalkohol
3-Chlorbenzylalkohol
Formel: C₇H₇ClO
Molecular Weight: 142.58 g/mol
Siedepunkt: 237
Dichte: 1.21 g/cm³ (20 °C)
MDL: MFCD00004632
CAS-Nummer: 873-63-2
EINECS: 212-847-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR