3-Chlor-p-kresol
Lieferant: CHEMPUR
Synonyme:
2-Chlor-4-hydroxytoluol, 3-Chlor-4-methylphenol, 1-Chlor-3-hydroxy-6-methylbenzol
FL18012.POR
018012-5G
018012-25G
CHMPFL18012.POREA
0
EUR
CHMPFL18012.POR
FLUO018012-5G
FLUO018012-25G
3-Chlor-p-kresol
3-Chlor-p-kresol
Formel:
C₇H₇ClO Molecular Weight: 142,58 g/mol |
MDL:
MFCD00060319 CAS-Nummer: 615-62-3 EINECS: 210-439-1 UN: 3437 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...