4'-Aminoacetophenon 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: p-Aminoacetophenon, 1-(4-Aminophenyl)ethanon

A13213.0B A13213.14 A13213.18 A13213.30
A13213.0BEA 533 EUR
A13213.0B A13213.14 A13213.18 A13213.30
4'-Aminoacetophenon 99%
4'-Aminoacetophenon

Purity: 99% CAS No.: 99-92-3 Molecular Formula: C8H9NO Formula Weight: 135.17 Melting Point: 103-108° Boiling Point: 292-294° Merck Reference: 14,413 MDL No.: MFCD00007896

Formel: C₈H₉NO
Molecular Weight: 135,17 g/mol
Siedepunkt: 292…294 °C
Schmelzpunkt: 103…108 °C
Dichte: 1,13 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007896
CAS-Nummer: 99-92-3
EINECS: 202-801-2
Merck Index: 14,413

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR