3'-Nitroacetophenon ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: m-Nitroacetophenone

A12155.0B A12155.30 A12155.36
A12155.0BEA 136 EUR
A12155.0B A12155.30 A12155.36
3'-Nitroacetophenon ≥98%
3'-Nitroacetophenon

Purity: 98+% CAS No.: 121-89-1 Molecular Formula: C8H7NO3 Formula Weight: 165.15 Odour: Slightly sweetish Melting Point: 76-80° Boiling Point: 202° Beilstein Reference: 743002 MDL No.: MFCD00007259

Formel: C₈H₇NO₃
Molecular Weight: 165.15 g/mol
Siedepunkt: 202 °C
Schmelzpunkt: 76…80 °C
Dichte: 1.433 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007259
CAS-Nummer: 121-89-1
EINECS: 204-504-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR