3',5'-Dimethoxyacetophenon
Lieferant: FLUOROCHEM
Synonyme:
1-(3,5-Dimethoxyphenyl)ethan-1-on
021803-5G
021803-25G
FLUO021803-5GEA
411
EUR
FLUO021803-5G
FLUO021803-25G
3',5'-Dimethoxyacetophenon
3',5'-Dimethoxyacetophenon
Formel:
C₁₀H₁₂O₃ Molecular Weight: 180,2 g/mol Siedepunkt: 290 °C (1013 hPa) Schmelzpunkt: 40…41 °C |
MDL:
MFCD00008739 CAS-Nummer: 39151-19-4 EINECS: 254-322-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...