2',5'-Dimethoxyacetophenon ≥99%

Lieferant: Thermo Scientific

Synonyme: 1-(2,5-Dimethoxyphenyl)ethan-1-on

A14973.06 A14973.14
A14973.06EA 23.1 EUR
A14973.06 A14973.14
2',5'-Dimethoxyacetophenon ≥99%
2',5'-Dimethoxyacetophenon

Purity: 99% CAS No.: 1201-38-3 Molecular Formula: C10H12O3 Formula Weight: 180.20 Flash Point: 110°(230°F) Refractive Index: 1.5435 Odour: Characteristic Melting Point: 18-20° Boiling Point: 154-156°/10mm Beilstein Reference: 2047423 MDL No.: MFCD00008728

Formel: C₁₀H₁₂O₃
Molecular Weight: 180,2 g/mol
Siedepunkt: 154…156 °C (10 mmHg)
Schmelzpunkt: 18…20 °C
Dichte: 1,139
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008728
CAS-Nummer: 1201-38-3
EINECS: 214-858-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR