4'-Hydroxypropiophenon ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
p-Hydroxypropiophenon, Paroxypropion
A15314.22
A15314.36
A15314.22EA
34.9
EUR
A15314.22
A15314.36
4'-Hydroxypropiophenon ≥99%
4'-Hydroxypropiophenon
Purity: 99%
CAS No.: 70-70-2
Molecular Formula: C9H10O2
Formula Weight: 150.18
Flash Point: 180°(356°F)
Melting Point: 148-151°
Beilstein Reference: 907511
Merck Reference: 14,7044
MDL No.: MFCD00002361
Formel:
C₉H₁₀O₂ Molecular Weight: 150.18 g/mol Siedepunkt: 140…145 °C (0.5 mmHg) Schmelzpunkt: 148…151 °C Dichte: 1.09 g/cm³ (20 °C) Flammpunkt: 180 °C (356 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002361 CAS-Nummer: 70-70-2 EINECS: 200-743-2 Merck Index: 12,07176 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...