2',4'-Dihydroxyacetophenon ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 1-(2,4-Dihydroxyphenyl)ethan-1-on

A12066.18 A12066.30
A12066.18EA 39.4 EUR
A12066.18 A12066.30
2',4'-Dihydroxyacetophenon ≥98%
2',4'-Dihydroxyacetophenon

Purity: 98% CAS No.: 89-84-9 Molecular Formula: C8H8O3 Formula Weight: 152.15 Melting Point: 142-146° Beilstein Reference: 1282505 Merck Reference: 14,8140 MDL No.: MFCD00002279

Formel: C₈H₈O₃
Molecular Weight: 152,15 g/mol
Siedepunkt: 336 °C (1013 hPa)
Schmelzpunkt: 142…146 °C
Dichte: 1,18 g/cm³ (141 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002279
CAS-Nummer: 89-84-9
EINECS: 201-945-3
Merck Index: 14,8140

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR