4-tert-Butylbenzoylchlorid ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
4-tert-Butylbenzoesäurechlorid
A14373.14
A14373.22
A14373.36
A14373.14EA
45.5
EUR
A14373.14
A14373.22
A14373.36
4-tert-Butylbenzoylchlorid ≥98%
4-tert-Butylbenzoylchlorid
Purity: 98%
CAS No.: 1710-98-1
Molecular Formula: C11H13ClO
Formula Weight: 196.68
Flash Point: 87°(188°F)
Refractive Index: 1.5351
Boiling Point: 125-126°/10mm
Beilstein Reference: 775793
UN No.: 3265
MDL No.: MFCD00000695
Formel:
C₁₁H₁₃ClO Molecular Weight: 196,68 g/mol Siedepunkt: 125…126 °C (10 mmHg) Dichte: 1,080 Flammpunkt: 87 °C (188 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000695 CAS-Nummer: 1710-98-1 EINECS: 216-973-1 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...